3,3-Dimethylglutaric anhydride ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4,4-Dimethyldihydro-2H-pyran-2,6(3H)-dione
D1323-25G
D1323-5G
TCIAD1323-25GEA
1610
DKK
TCIAD1323-25G
TCIAD1323-5G
3,3-Dimethylglutaric anhydride ≥97.0% (by titrimetric analysis)
3,3-Dimethylglutaric anhydride
Formel:
C₇H₁₀O₃ |
MDL Number:
MFCD00006684 CAS-nummer: 4160-82-1 EINECS: 223-998-1 |
|
Specifikationer Testresultater
Appearance | White powder to crystal |
Purity (Morpholine Method) | min. 97.0 % |
Melting point | 123.0 to 127.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...