Betaine hydrochloride ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B0458-25G
B0458-500G
TCIAB0458-25GEA
337
DKK
TCIAB0458-25G
TCIAB0458-500G
Betaine hydrochloride ≥98.0% (by titrimetric analysis)
Betaine hydrochloride
Formel:
C₅H₁₂ClNO₂ Molekylvægt: 153.61 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011903 CAS-nummer: 590-46-5 EINECS: 209-683-1 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Suitability for protein-analysis | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...