Ampicillin trihydrat ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate

A2092-25G A2092-5G
TCIAA2092-25GEA 370 DKK
TCIAA2092-25G TCIAA2092-5G
Ampicillin trihydrat ≥98.0% (by HPLC, titration analysis)
Ampicillin trihydrat
Formel: C₁₆H₁₉N₃O₄S·3H₂O
Smeltepunkt: 202°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00072036
CAS-nummer: 7177-48-2
EINECS: 200-709-7
Merck Index: 14,00586
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4f7e951dc4b19e89ce51e87ebdfcb5bb39c6f1e&vid=d4f7e951dc4b19e89ce51e87ebdfcb5bb39c6f1e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +280.0 to +305.0 deg (C=0.5, H2O) (calcd.on anh.substance)
Water 12.0 to 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR