S-Ethylisothiouroniumbromid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Ethyl carbamimidothioate hydrobromide, S-Ethylisothiourea Hydrobromide, S-EIT hydrobromide, 2-Ethyl-2-thiopseudourea hydrobromide
E0182-25G
E0182-100G
TCIAE0182-25GEA
624
DKK
TCIAE0182-25G
TCIAE0182-100G
S-Ethylisothiouroniumbromid ≥98.0% (by titrimetric analysis)
S-Ethylisothiouronium bromide
Formel:
C₃H₉BrN₂S |
MDL Number:
MFCD00012585 CAS-nummer: 1071-37-0 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...