Abietinsyre ≥80.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A0001-500G
A0001-25G
TCIAA0001-500GEA
1420
DKK
TCIAA0001-500G
TCIAA0001-25G
Abietinsyre ≥80.0% (by GC)
Abietinsyre
Formel:
C₂₀H₃₀O₂ |
MDL Number:
MFCD03423567 CAS-nummer: 514-10-3 EINECS: 208-178-3 UN nummer: 3077 ADR: 9,III Merck Index: 14,00007 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 80.0 % |
Melting point | 159.0 to 169.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...