N-Benzylglycinethylester ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B2273-25G B2273-5G
TCIAB2273-25GEA 632 DKK
TCIAB2273-25G TCIAB2273-5G
N-Benzylglycinethylester ≥96.0% (by GC)
N-Benzylglycinethylester
Formel: C₁₁H₁₅NO₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00009174
CAS-nummer: 6436-90-4
EINECS: 229-218-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ae5377502218b2311102b75c86e2ffb255e34b&vid=c9ae5377502218b2311102b75c86e2ffb255e34b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR