Pyrrole-2-carboxylic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
P1270-1G
P1270-5G
TCIAP1270-1GEA
132
DKK
TCIAP1270-1G
TCIAP1270-5G
Pyrrole-2-carboxylic acid ≥98.0% (by titrimetric analysis)
Pyrrole-2-carboxylic acid
Formel:
C₅H₅NO₂ Molekylvægt: 111.10 g/mol |
MDL Number:
MFCD00005219 CAS-nummer: 634-97-9 EINECS: 211-221-9 |
|
Specifikationer Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...