Glycylglycylglycine (Gly-Gly-Gly)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

G0126-25G G0126-5G
TCIAG0126-25GEA 1560 DKK
TCIAG0126-25G TCIAG0126-5G
Glycylglycylglycine (Gly-Gly-Gly)
Proteiner

  • EINECS: 209-122-0
  • Solubility in water: Almost transparency
  • Product Categories: Bioscience, Oligopeptides
  • Other Product Names/Synonyms: H-Gly-Gly-Gly-OH, Triglycine

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=565fef6a8b417bd6f4e4c6dfa71027075a9fc939&vid=e92013f200d861f6a75a5724cb5aa66ca3f5f1a7&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR