Monuron (N-(4-Chlorphenyl)-N',N'-dimethylcarbamid) ≥95.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C0254-25G C0254-500G
TCIAC0254-25GEA 165 DKK
TCIAC0254-25G TCIAC0254-500G
Monuron (N-(4-Chlorphenyl)-N',N'-dimethylcarbamid) ≥95.0% (by HPLC, total nitrogen)
Monuron (N-(4-Chlorphenyl)-N',N'-dimethylcarbamid)
Formel: C₉H₁₁ClN₂O
MDL Number: MFCD00018556
CAS-nummer: 150-68-5
EINECS: 205-766-1
UN nummer: 3077
ADR: 9,III
Merck Index: 14,06261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=974bfc7f1451e5606f158dacfd885c7ce90b3c53&vid=974bfc7f1451e5606f158dacfd885c7ce90b3c53&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 172.0 to 177.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR