Tris(2-aminoethyl)amin ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
N1,N1-Bis(2-aminoethyl)ethane-1,2-diamine, TREN, 2,2',2''-Triaminotriethylamine
T1243-25ML
T1243-5ML
T1243-100ML
TCIAT1243-25MLEA
796
DKK
TCIAT1243-25ML
TCIAT1243-5ML
TCIAT1243-100ML
Tris(2-aminoethyl)amin ≥98.0% (by titrimetric analysis)
Tris(2-aminoethyl)amin
Formel:
C₆H₁₈N₄ Kogepunkt: 114°C |
MDL Number:
MFCD00008177 CAS-nummer: 4097-89-6 EINECS: 223-857-4 UN nummer: 2922 ADR: 8,II |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...