Arecoliniumbromid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Arecoline hydrobromide

A0523-25G A0523-5G
TCIAA0523-25GEA 1900 DKK
TCIAA0523-25G TCIAA0523-5G
Arecoliniumbromid ≥98.0% (by HPLC, titration analysis)
Arecoliniumbromid
Formel: C₈H₁₄BrNO₂
MDL Number: MFCD00039041
CAS-nummer: 300-08-3
EINECS: 206-087-3
UN nummer: 1544
ADR: 6.1,III
Merck Index: 14,00778
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bea17e6f17e13cd1916e8eda9f10a23d0f14b4a&vid=1bea17e6f17e13cd1916e8eda9f10a23d0f14b4a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 169.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR