2,4,6-Trimethoxypyrimidin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T2039-1G T2039-5G
TCIAT2039-1GEA 378 DKK
TCIAT2039-1G TCIAT2039-5G
2,4,6-Trimethoxypyrimidin ≥98.0% (by GC)
2,4,6-Trimethoxypyrimidine
Formel: C₇H₁₀N₂O₃
Kogepunkt: 232°C
MDL Number: MFCD00829225
CAS-nummer: 13106-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96314f723a5d94435384ee98bf9975dabaf86768&vid=96314f723a5d94435384ee98bf9975dabaf86768&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR