2-Brom-9,10-anthraquinon ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
2-Bromoanthracene-9,10-dione, 2-Bromo-9,10-anthracenedione
B2689-1G
B2689-25G
B2689-5G
TCIAB2689-1GEA
485
DKK
TCIAB2689-1G
TCIAB2689-25G
TCIAB2689-5G
2-Brom-9,10-anthraquinon ≥96.0% (by GC)
2-Brom-9,10-anthraquinon
Formel:
C₁₄H₇BrO₂ Molekylvægt: 287.11 g/mol |
MDL Number:
MFCD00474541 CAS-nummer: 572-83-8 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 205.0 to 211.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...