Hexamethyldisilathian ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Bis(trimethylsilyl)sulphide

H0871-25G H0871-5G
TCIAH0871-25GEA 3020 DKK
TCIAH0871-25G TCIAH0871-5G
Hexamethyldisilathian ≥97.0% (by GC)
Hexamethyldisilathian
Formel: C₆H₁₈SSi₂
Molekylvægt: 178.45 g/mol
Kogepunkt: 163°C
Flammepunkt: 23°C
MDL Number: MFCD00014851
CAS-nummer: 3385-94-2
EINECS: 222-201-4
UN nummer: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc31166238c9427eea5b66a3f5f5015b021911&vid=13cc31166238c9427eea5b66a3f5f5015b021911&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR