Di-tert-butylsilan ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

D3731-1G D3731-5G
TCIAD3731-1GEA 591 DKK
TCIAD3731-1G TCIAD3731-5G
Di-tert-butylsilan ≥95.0% (by GC)
Di-tert-butylsilan
Formel: C₈H₂₀Si
Kogepunkt: 129°C
Flammepunkt: –2°C
MDL Number: MFCD00270996
CAS-nummer: 30736-07-3
UN nummer: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18ae86af26a4186aa621c9e7657d181ac18fba6c&vid=18ae86af26a4186aa621c9e7657d181ac18fba6c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR