Rhodanin-N-eddikesyre ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(4-Oxo-2-thioxothiazolidin-3-yl)acetic acid, Rhodanine-3-acetic acid

R0054-25G R0054-5G
TCIAR0054-25GEA 419 DKK
TCIAR0054-25G TCIAR0054-5G
Rhodanin-N-eddikesyre ≥98.0% (by titrimetric analysis)
Rhodanin-N-eddikesyre
Formel: C₅H₅NO₃S₂
MDL Number: MFCD00005491
CAS-nummer: 5718-83-2
EINECS: 227-220-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=faa755606ecdd76c3e32891bb8129f89dc6ea3c2&vid=faa755606ecdd76c3e32891bb8129f89dc6ea3c2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light red to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 145.0 to 149.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR