5-Methylthiophen-2-carboxaldehyd ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Methylthiophene-2-Carbaldehyde, 5-Methyl-2-thiophenecarboxaldehyde, 5-Methylthiophene-2-carbaldehyde
M0911-5G
M0911-25G
TCIAM0911-5GEA
140
DKK
TCIAM0911-5G
TCIAM0911-25G
5-Methylthiophen-2-carboxaldehyd ≥97.0% (by GC)
5-Methylthiophen-2-carboxaldehyd
Formel:
C₆H₆OS Kogepunkt: 205°C Flammepunkt: 87°C |
MDL Number:
MFCD00005434 CAS-nummer: 13679-70-4 EINECS: 237-178-6 |
|
Specifikationer Testresultater
Appearance | Yellow to Amber to Dark purple clear liquid to cloudy liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...