O-Pentafluorphenyl chlorthioformat ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: O-Perfluorophenyl chlorothioformate, Chlorothioformic acid O-Pentafluorophenyl ester, Pentafluorophenyl chlorothionoformate, Pentafluorophenyl Thionochloroformate

P1267-1G P1267-5G
TCIAP1267-1GEA 362 DKK
TCIAP1267-1G TCIAP1267-5G
O-Pentafluorphenyl chlorthioformat ≥95.0% (by GC)
O-Pentafluorphenyl chlorthioformat
Formel: C₇ClF₅OS
Flammepunkt: 87°C
MDL Number: MFCD00075405
CAS-nummer: 135192-53-9
UN nummer: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ead08ecc8cb01b9eaf0e39f73dc4c89c03419658&vid=ead08ecc8cb01b9eaf0e39f73dc4c89c03419658&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR