2-(2-Bromethyl)-1,3-dioxolan ≥90.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B1132-25G B1132-100G
TCIAB1132-25GEA 657 DKK
TCIAB1132-25G TCIAB1132-100G
2-(2-Bromethyl)-1,3-dioxolan ≥90.0% (by GC)
2-(2-Bromethyl)-1,3-dioxolan
Formel: C₅H₉BrO₂
Molekylvægt: 181.03 g/mol
Kogepunkt: 73°C
Flammepunkt: 65°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00003216
CAS-nummer: 18742-02-4
EINECS: 242-551-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5d4d65e5849014140f1412c84e03b130a94bbb4&vid=f5d4d65e5849014140f1412c84e03b130a94bbb4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR