1-Phenyl-3-pyrazolidon ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Phenyl-3-pyrazolidinone

P0153-25G P0153-500G
TCIAP0153-25GEA 140 DKK
TCIAP0153-25G TCIAP0153-500G
1-Phenyl-3-pyrazolidon ≥98.0% (by titrimetric analysis)
1-Phenyl-3-pyrazolidon
Formel: C₉H₁₀N₂O
MDL Number: MFCD00003094
CAS-nummer: 92-43-3
EINECS: 202-155-1
UN nummer: 2811
ADR: 6.1,III
Merck Index: 14,07309
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0f9f805f59ac8df6df54e3a82a8f81903211623&vid=f0f9f805f59ac8df6df54e3a82a8f81903211623&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 121.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR