3,3-Dimethylbutyraldehyd ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
3,3-Dimethylbutanal
D4098-25ML
D4098-5ML
TCIAD4098-25MLEA
1400
DKK
TCIAD4098-25ML
TCIAD4098-5ML
3,3-Dimethylbutyraldehyd ≥97.0% (by GC)
3,3-Dimethylbutyraldehyd
Formel:
C₆H₁₂O Molekylvægt: 100.16 g/mol Kogepunkt: 104°C Flammepunkt: 13°C |
MDL Number:
MFCD00042807 CAS-nummer: 2987-16-8 EINECS: 221-054-3 UN nummer: 1989 ADR: 3 II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Specific gravity (20/20) | 0.8000 to 0.8100 |
Refractive index n20/D | 1.3950 to 1.3990 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...