2-(Ethylthio)ethanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethyl 2-hydroxyethyl sulphide, 2-(Ethylmercapto)-ethanol

E0248-25ML E0248-500ML E0248-100ML
TCIAE0248-25MLEA 132 DKK
TCIAE0248-25ML TCIAE0248-500ML TCIAE0248-100ML
2-(Ethylthio)ethanol ≥98.0% (by GC)
2-(Ethylthio)ethanol
Formel: C₄H₁₀OS
Molekylvægt: 106.19 g/mol
Kogepunkt: 182°C
Flammepunkt: 93°C
MDL Number: MFCD00002909
CAS-nummer: 110-77-0
EINECS: 203-802-0
UN nummer: 1760
ADR: 8 III
Merck Index: 14,03857
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75ab1f71099c9cf2f1187a010fc1baf606e2f761&vid=75ab1f71099c9cf2f1187a010fc1baf606e2f761&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR