2-Hydroxyethylsalicylat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

E0113-25G E0113-500G
TCIAE0113-25GEA 140 DKK
TCIAE0113-25G TCIAE0113-500G
2-Hydroxyethylsalicylat ≥98.0% (by GC)
2-Hydroxyethylsalicylat
Formel: C₉H₁₀O₄
Molekylvægt: 182.18 g/mol
Kogepunkt: 172°C
MDL Number: MFCD00002862
CAS-nummer: 87-28-5
EINECS: 201-737-2
Merck Index: 14,04499
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e24d907a6e15fb6cdc16ab9fa0384ecfdf4c1623&vid=e24d907a6e15fb6cdc16ab9fa0384ecfdf4c1623&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR