Methylmercaptoacetat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methyl thioglycolate

T1287-25G T1287-500G T1287-100G
TCIAT1287-25GEA 132 DKK
TCIAT1287-25G TCIAT1287-500G TCIAT1287-100G
Methylmercaptoacetat ≥98.0% (by GC)
Methylmercaptoacetat
Formel: C₃H₆O₂S
Molekylvægt: 106.15 g/mol
Kogepunkt: 49°C
Flammepunkt: 51°C
MDL Number: MFCD00004873
CAS-nummer: 2365-48-2
EINECS: 219-121-7
UN nummer: 1992
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcb2558fd8203305953102c31b1f6cd95f1a2a92&vid=bcb2558fd8203305953102c31b1f6cd95f1a2a92&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR