9-Acridone ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 9,10-Dihydro-9-oxoacridin, 9(10H)-Acridanone, Acridone, 9(10H)-Acridone, Acridin-9(10H)-one, 9-Acridone

A0133-25G A0133-5G
TCIAA0133-25GEA 1290 DKK
TCIAA0133-25G TCIAA0133-5G
9-Acridone ≥98.0% (by HPLC, total nitrogen)
9-Acridone
Formel: C₁₃H₉NO
Molekylvægt: 195.22 g/mol
Smeltepunkt: 354°C
MDL Number: MFCD00005019
CAS-nummer: 578-95-0
EINECS: 209-434-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e5418d3b41c02d63ead8fb883be41ec96e4a59&vid=a0e5418d3b41c02d63ead8fb883be41ec96e4a59&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR