2,2-Bis(hydroxymethyl)propionsyre ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid

B1192-25G B1192-500G
TCIAB1192-25GEA 132 DKK
TCIAB1192-25G TCIAB1192-500G
2,2-Bis(hydroxymethyl)propionsyre ≥97.0% (by GC, titration analysis)
2,2-Bis(hydroxymethyl)propionsyre
Formel: C₅H₁₀O₄
Molekylvægt: 134.13 g/mol
MDL Number: MFCD00004199
CAS-nummer: 4767-03-7
EINECS: 225-306-3
Merck Index: 14,03253
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0004a5d336a9c664442eeab33c0677777a294ac8&vid=0004a5d336a9c664442eeab33c0677777a294ac8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 179.0 to 187.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR