Ethyl oxalylchlorid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethyl oxalyl monochloride, Oxalic Acid Monoethyl Ester Chloride, Ethyl Chlorooxoacetate, Ethyl Chloroglyoxylate, Chloroglyoxylic Acid Ethyl Ester, Ethyl chloroformylformate

C1066-25G C1066-500G C1066-100G
TCIAC1066-25GEA 288 DKK
TCIAC1066-25G TCIAC1066-500G TCIAC1066-100G
Ethyl oxalylchlorid ≥98.0% (by titrimetric analysis)
Ethyl oxalylchlorid
Formel: C₄H₅ClO₃
Flammepunkt: 41°C
MDL Number: MFCD00000706
CAS-nummer: 4755-77-5
EINECS: 225-285-0
UN nummer: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cc33659c5e69bce1ec23eb9b6e8664d04c7bbe4&vid=0cc33659c5e69bce1ec23eb9b6e8664d04c7bbe4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR