Ethyl 3,3-diethoxypropionat 95,0 GC_ASSAY_METHOD

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ethyl 3,3-diethoxypropanoate, Ethyl 3,3-diethoxypropionate

D1962-100ML D1962-25ML
TCIAD1962-100MLEA 1250 DKK
TCIAD1962-100ML TCIAD1962-25ML
Ethyl 3,3-diethoxypropionat 95,0 GC_ASSAY_METHOD
Ethyl 3,3-diethoxypropionat
Formel: C₉H₁₈O₄
Molekylvægt: 190.24 g/mol
Kogepunkt: 86°C
Flammepunkt: 84°C
MDL Number: MFCD00009865
CAS-nummer: 10601-80-6
EINECS: 234-223-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7ecaa3892edd4db29a819a823fae8c66a9433c9&vid=b7ecaa3892edd4db29a819a823fae8c66a9433c9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR