Adamantan ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A0696-25G A0696-500G
TCIAA0696-25GEA 288 DKK
TCIAA0696-25G TCIAA0696-500G
Adamantan ≥99.0% (by GC)
Adamantan
Formel: C₁₀H₁₆
Kogepunkt: 171 °C (1013 hPa)
Smeltepunkt: –25 °C
Massefylde: 1,07 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00074719
CAS-nummer: 281-23-2
EINECS: 206-001-4
Merck Index: 14,00149
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e15f1504c72fba7b59c0633bd0509a489de4ded3&vid=e15f1504c72fba7b59c0633bd0509a489de4ded3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR