4-Phenylpyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
P0162-25G
P0162-5G
TCIAP0162-25GEA
1230
DKK
TCIAP0162-25G
TCIAP0162-5G
4-Phenylpyridin ≥98.0% (by GC)
4-Phenylpyridine
Formel:
C₁₁H₉N Kogepunkt: 281°C Smeltepunkt: 78 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006420 CAS-nummer: 939-23-1 EINECS: 213-357-4 |
|
Specifikationer Testresultater
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 74.0 to 77.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...