3-Phenylpyridin ≥94.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

P1040-25G P1040-5G
TCIAP1040-25GEA 1680 DKK
TCIAP1040-25G TCIAP1040-5G
3-Phenylpyridin ≥94.0% (by GC)
3-Phenylpyridine
Formel: C₁₁H₉N
Kogepunkt: 274°C
Smeltepunkt: 76…77 °C
Massefylde: 1,08 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006380
CAS-nummer: 1008-88-4
EINECS: 213-762-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c90e12147a7d573cfc04803e514b66a3c3093078&vid=c90e12147a7d573cfc04803e514b66a3c3093078&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR