2-Phenylpyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
P1039-25G
P1039-5G
TCIAP1039-25GEA
1050
DKK
TCIAP1039-25G
TCIAP1039-5G
2-Phenylpyridin ≥98.0% (by GC)
2-Phenylpyridine
Formel:
C₁₁H₉N Kogepunkt: 270°C Smeltepunkt: 76…77 °C |
MDL Number:
MFCD00006280 CAS-nummer: 1008-89-5 EINECS: 213-763-1 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...