3-Methyl-2-phenylpyridin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Phenyl-beta-picoline

M0932-25G M0932-5G
TCIAM0932-25GEA 3760 DKK
TCIAM0932-25G TCIAM0932-5G
3-Methyl-2-phenylpyridin ≥98.0% (by GC)
3-Methyl-2-phenylpyridine
Formel: C₁₂H₁₁N
Kogepunkt: 160°C
Massefylde: 1,0648 g/cm³ (25 °C)
Flammepunkt: 96°C
MDL Number: MFCD00006283
CAS-nummer: 10273-90-2
EINECS: 233-619-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb8d82c9b5dcd9937e234da850b8e400a4dae186&vid=cb8d82c9b5dcd9937e234da850b8e400a4dae186&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR