2-Chlor-3-methyl-5-nitropyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Chloro-5-nitro-3-picoline
C2020-1G
C2020-5G
TCIAC2020-1GEA
197
DKK
TCIAC2020-1G
TCIAC2020-5G
2-Chlor-3-methyl-5-nitropyridin ≥98.0% (by GC)
2-Chloro-3-methyl-5-nitropyridine
Formel:
C₆H₅ClN₂O₂ Molekylvægt: 172.57 g/mol Kogepunkt: 145°C |
MDL Number:
MFCD00173686 CAS-nummer: 22280-56-4 EINECS: 244-889-5 |
|
Specifikationer Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 47.0 to 51.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...