2-Aminopyridin ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Pyridylamine, Pyridin-2-amine

A0411-25G A0411-500G A0411-100G
TCIAA0411-25GEA 132 DKK
TCIAA0411-25G TCIAA0411-500G TCIAA0411-100G
2-Aminopyridin ≥99.0% (by GC, titration analysis)
2-Aminopyridine
Formel: C₅H₆N₂
Kogepunkt: 105°C
Smeltepunkt: 50…52 °C
Massefylde: 1,24 g/cm³ (20 °C)
Flammepunkt: 92 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00006312
CAS-nummer: 504-29-0
EINECS: 207-988-4
UN nummer: 2671
ADR: 6.1,II
Merck Index: 14,00473
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3822a3ec11cdee4f04df5e5a1de2399015efc8d3&vid=3822a3ec11cdee4f04df5e5a1de2399015efc8d3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 57.0 to 61.0 deg-C
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR