2-Fluorpyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
F0217-25G
F0217-100G
TCIAF0217-25GEA
296
DKK
TCIAF0217-25G
TCIAF0217-100G
2-Fluorpyridin ≥98.0% (by GC)
2-Fluoropyridine
Formel:
C₅H₄FN Kogepunkt: 126°C Massefylde: 1,129 g/cm³ (20 °C) Flammepunkt: 24°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006224 CAS-nummer: 372-48-5 EINECS: 206-757-5 UN nummer: 1993 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...