2-Chlor-6-methoxypyridin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C0197-25G C0197-5G
TCIAC0197-25GEA 542 DKK
TCIAC0197-25G TCIAC0197-5G
2-Chlor-6-methoxypyridin ≥98.0% (by GC)
2-Chloro-6-methoxypyridine
Formel: C₆H₆ClNO
Kogepunkt: 184°C
Massefylde: 1,207 g/cm³ (20 °C)
Flammepunkt: 76°C
MDL Number: MFCD00006265
CAS-nummer: 17228-64-7
EINECS: 241-264-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d443d702897288f81574adbcb25c385a02ec9db&vid=8d443d702897288f81574adbcb25c385a02ec9db&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR