2-Chlor-5-nitropyridin ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C0025-25G C0025-5G
TCIAC0025-25GEA 501 DKK
TCIAC0025-25G TCIAC0025-5G
2-Chlor-5-nitropyridin ≥99.0% (by GC)
2-Chloro-5-nitropyridine
Formel: C₅H₃ClN₂O₂
Smeltepunkt: 107 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006240
CAS-nummer: 4548-45-2
EINECS: 224-908-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77688160bcf8cdba3f488f7bc9515a7e01e7e996&vid=77688160bcf8cdba3f488f7bc9515a7e01e7e996&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 107.0 to 111.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR