3-Brom-2-chlorpyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B2328-25G
B2328-5G
TCIAB2328-25GEA
559
DKK
TCIAB2328-25G
TCIAB2328-5G
3-Brom-2-chlorpyridin ≥98.0% (by GC)
3-Bromo-2-chloropyridine
Formel:
C₅H₃BrClN Kogepunkt: 97°C Smeltepunkt: 55 °C |
MDL Number:
MFCD00234007 CAS-nummer: 52200-48-3 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 59.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...