3,5-Dibrompyridin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D2573-25G D2573-5G
TCIAD2573-25GEA 1010 DKK
TCIAD2573-25G TCIAD2573-5G
3,5-Dibrompyridin ≥98.0% (by GC)
3,5-Dibromopyridine
Formel: C₅H₃Br₂N
Kogepunkt: 222°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00014634
CAS-nummer: 625-92-3
EINECS: 210-916-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96fb32c9d59677ea2eaeea566927907760ccc64a&vid=96fb32c9d59677ea2eaeea566927907760ccc64a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in Toluene very faint turbidity
Melting point 111.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR