2-Naphtol ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: β-Naphtol

N0027-25G N0027-250G
TCIAN0027-25GEA 132 DKK
TCIAN0027-25G TCIAN0027-250G
2-Naphtol ≥99.0% (by GC)
2-Naphtol
Formel: C₁₀H₈O
Kogepunkt: 286 °C (1013 hPa)
Massefylde: 1,217 g/cm³ (20 °C)
Flammepunkt: 153 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004067
CAS-nummer: 135-19-3
EINECS: 205-182-7
ADR: 9,III
Merck Index: 14,06384
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39fd47b639448a316bca5603e72136b283ea910c&vid=39fd47b639448a316bca5603e72136b283ea910c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 120.0 to 124.0 deg-C
Solubility in EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR