Isochinolin ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

I0182-25G I0182-500G
TCIAI0182-25GEA 132 DKK
TCIAI0182-25G TCIAI0182-500G
Isochinolin ≥95.0% (by GC)
Isoquinoline
Formel: C₉H₇N
Molekylvægt: 129.16 g/mol
Kogepunkt: 242°C
Flammepunkt: 107
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00006898
CAS-nummer: 119-65-3
EINECS: 204-341-8
UN nummer: 2811
ADR: 6.1 III
Merck Index: 14,05222
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b410af3ccc5fa5716763526b44a7f5b34554bf16&vid=b410af3ccc5fa5716763526b44a7f5b34554bf16&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR