5-Nitroisoquinolin
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
N0640-25G
N0640-5G
TCIAN0640-25GEA
1020
DKK
TCIAN0640-25G
TCIAN0640-5G
5-Nitroisoquinolin
5-Nitroisoquinolin
Formel:
C₉H₆N₂O₂ Kogepunkt: 340 °C (1013 hPa) Smeltepunkt: 111…113 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006905 CAS-nummer: 607-32-9 EINECS: 210-133-8 |
|
Specifikationer Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 106.0 to 115.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...