5-Brom-2-nitropyridin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B2766-1G B2766-5G
TCIAB2766-1GEA 132 DKK
TCIAB2766-1G TCIAB2766-5G
5-Brom-2-nitropyridin ≥98.0% (by GC)
5-Brom-2-nitropyridin
Formel: C₅H₃BrN₂O₂
MDL Number: MFCD00160411
CAS-nummer: 39856-50-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d54649c2362121a028459f87e867adec5dc6a1d2&vid=d54649c2362121a028459f87e867adec5dc6a1d2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 148.0 to 152.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR