Methyl6-aminonicotinat ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 6-aminonicotinate, 6-Aminonicotinic acid methyl ester

M2107-25G M2107-5G
TCIAM2107-25GEA 2380 DKK
TCIAM2107-25G TCIAM2107-5G
Methyl6-aminonicotinat ≥98.0% (by GC, titration analysis)
Methyl6-aminonicotinat
Formel: C₇H₈N₂O₂
Molekylvægt: 152.15 g/mol
MDL Number: MFCD00797844
CAS-nummer: 36052-24-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f31af78813a7f0cfbe7804b348656dec990009e2&vid=f31af78813a7f0cfbe7804b348656dec990009e2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 158.0 to 162.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR