Methyl2-chlornicotinat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M2165-25G M2165-5G
TCIAM2165-25GEA 780 DKK
TCIAM2165-25G TCIAM2165-5G
Methyl2-chlornicotinat ≥98.0% (by GC)
Methyl2-chlornicotinat
Formel: C₇H₆ClNO₂
Kogepunkt: 70°C
MDL Number: MFCD01318629
CAS-nummer: 40134-18-7
EINECS: 254-805-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b442c531c7680583fdcbb811832af86a88cbb892&vid=b442c531c7680583fdcbb811832af86a88cbb892&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR