Methyl5-Bromnicotinat ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 5-bromonicotinate

M2008-1G M2008-5G
TCIAM2008-1GEA 132 DKK
TCIAM2008-1G TCIAM2008-5G
Methyl5-Bromnicotinat ≥98.0% (by GC, titration analysis)
Methyl5-Bromnicotinat
Formel: C₇H₆BrNO₂
Molekylvægt: 216.03 g/mol
MDL Number: MFCD00173668
CAS-nummer: 29681-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=605ae8ca4b425c71af10d9898db33f5fc1387b2f&vid=605ae8ca4b425c71af10d9898db33f5fc1387b2f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR