Aminoguanidinium hydrogencarbonat ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Aminoguanidine hydrogen carbonate, Aminoguanidine bicarbonate, 1-Aminoguanidinium hydrogen carbonate, Aminoguanidine hydrogencarbonate, Guanylhydrazine hydrogencarbonate

A0307-25G A0307-500G
TCIAA0307-25GEA 148 DKK
TCIAA0307-25G TCIAA0307-500G
Aminoguanidinium hydrogencarbonat ≥99.0% (by titrimetric analysis)
Aminoguanidinium hydrogencarbonat
Formel: C₂H₈N₄O₃
Molekylvægt: 136.11 g/mol
MDL Number: MFCD00012949
CAS-nummer: 2582-30-1
EINECS: 219-956-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59070d3eaa6bf67c4d79c92c62c73b02cc323a21&vid=59070d3eaa6bf67c4d79c92c62c73b02cc323a21&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Purity (Potassium iodate Method) min. 99.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR