Natrium-3-nitrobenzensulfonat ≥95.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Sodium 3-nitrobenzenesulphonate, 3-Nitrobenzenesulphonic acid, sodium salt
N0141-25G
N0141-500G
TCIAN0141-25GEA
132
DKK
TCIAN0141-25G
TCIAN0141-500G
Natrium-3-nitrobenzensulfonat ≥95.0% (by titrimetric analysis)
Natrium-3-nitrobenzensulfonat
Formel:
C₆H₄NNaO₅S Molekylvægt: 225.16 g/mol |
MDL Number:
MFCD00007490 CAS-nummer: 127-68-4 EINECS: 204-857-3 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 94.0 area% |
Purity (Ion exchange titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...