Acetylendicarboxylsyre ≥90.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A0088-25G A0088-5G
TCIAA0088-25GEA 1010 DKK
TCIAA0088-25G TCIAA0088-5G
Acetylendicarboxylsyre ≥90.0% (by HPLC)
Acetylendicarboxylsyre
Formel: C₄H₂O₄
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00004362
CAS-nummer: 142-45-0
EINECS: 205-536-0
UN nummer: 2923
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83b285b6720bc6dffc3a8ebcb323f729cf657384&vid=83b285b6720bc6dffc3a8ebcb323f729cf657384&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 90.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR